Pyridazine-4-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Formyl-1,2-diazine, 4-Formylpyridazine
OR59872-1G
OR59872-250MG
OR59872-5G
APOSOR59872-1GEA
317
EUR
APOSOR59872-1G
APOSOR59872-250MG
APOSOR59872-5G
Pyridazine-4-carboxaldehyde
Pyridazine-4-carboxaldehyde
Formel:
C₅H₄N₂O Molecular Weight: 108,098 g/mol Schmelzpunkt: 90…91 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06227447 CAS-Nummer: 50901-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...