(2-Methyl-1,3-thiazol-4-yl)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2-Methylthiazol-4-yl)acetic acid, 4-(Carboxymethyl)-2-methyl-1,3-thiazole
OR60043-1G
OR60043-250MG
APOSOR60043-1GEA
81.2
EUR
APOSOR60043-1G
APOSOR60043-250MG
(2-Methyl-1,3-thiazol-4-yl)acetic acid
(2-Methyl-1,3-thiazol-4-yl)acetic acid
Formel:
C₆H₇NO₂S Molecular Weight: 157,19 g/mol Schmelzpunkt: 123,5…124.5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02669593 CAS-Nummer: 13797-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...