7-Chloropyrido[3,4-b]pyrazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Chloro-6-azaquinoxaline

OR60108-1G OR60108-2.5G OR60108-500MG
APOSOR60108-1GEA 212 EUR
APOSOR60108-1G APOSOR60108-2.5G APOSOR60108-500MG
7-Chloropyrido[3,4-b]pyrazine
7-Chloropyrido[3,4-b]pyrazine
Formel: C₇H₄ClN₃
Molecular Weight: 165,58 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11044722
CAS-Nummer: 93049-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR