5-Chloro-2-methoxynicotinonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-3-cyano-2-methoxypyridine, 5-Chloro-2-methoxypyridine-3-carbonitrile

OR60154-1G OR60154-250MG
APOSOR60154-1GEA 248 EUR
APOSOR60154-1G APOSOR60154-250MG
5-Chloro-2-methoxynicotinonitrile
5-Chloro-2-methoxynicotinonitrile
Formel: C₇H₅ClN₂O
Molecular Weight: 168.58 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17214212
CAS-Nummer: 1256812-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR