7-Bromo-8-methylquinoline 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-8-methyl-1-azanaphthalene

OR60185-1G OR60185-250MG
APOSOR60185-1GEA 29 EUR
APOSOR60185-1G APOSOR60185-250MG
7-Bromo-8-methylquinoline 95+%
7-Bromo-8-methylquinoline
Formel: C₁₀H₈BrN
Molecular Weight: 222,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907849
CAS-Nummer: 809248-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR