8-Bromo-4-hydroxyquinoline 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR60188-1G
OR60188-5G
OR60188-25G
APOSOR60188-1GEA
43.5
EUR
APOSOR60188-1G
APOSOR60188-5G
APOSOR60188-25G
8-Bromo-4-hydroxyquinoline 95+%
8-Bromo-4-hydroxyquinoline
Formel:
C₉H₆BrNO Schmelzpunkt: 168…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00272389 CAS-Nummer: 57798-00-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...