6-Amino-4-bromoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-4-bromo-1-azanaphthalene, 4-Bromoquinolin-6-amine

OR60190-250MG OR60190-500MG OR60190-100MG
APOSOR60190-250MGEA 132 EUR
APOSOR60190-250MG APOSOR60190-500MG APOSOR60190-100MG
6-Amino-4-bromoquinoline
6-Amino-4-bromoquinoline
Formel: C₉H₇BrN₂
Molecular Weight: 223,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16659607
CAS-Nummer: 1260785-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR