5-Bromo-3-chloro-2-hydrazinopyrazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-chloro-2-hydrazino-1,4-diazine, (5-Bromo-3-chloropyrazin-2-yl)hydrazine

OR60219-1G OR60219-250MG OR60219-100MG
APOSOR60219-1GEA 401 EUR
APOSOR60219-1G APOSOR60219-250MG APOSOR60219-100MG
5-Bromo-3-chloro-2-hydrazinopyrazine
5-Bromo-3-chloro-2-hydrazinopyrazine
Formel: C₄H₄BrClN₄
Molecular Weight: 223,458 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22375535
CAS-Nummer: 850421-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR