5-Chloro-3-methylbenzofuran

Lieferant: Apollo Scientific
Achtung

OR60263-1G OR60263-250MG
APOSOR60263-1GEA 1060 EUR
APOSOR60263-1G APOSOR60263-250MG
5-Chloro-3-methylbenzofuran
5-Chloro-3-methylbenzofuran
Formel: C₉H₇ClO
Molecular Weight: 166.604 g/mol
Siedepunkt: 232…233 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797625
CAS-Nummer: 1125-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR