6-Chloro-3-methylbenzofuran

Lieferant: Apollo Scientific
Achtung

OR60264-1G OR60264-250MG
APOSOR60264-1GEA 986 EUR
APOSOR60264-1G APOSOR60264-250MG
6-Chloro-3-methylbenzofuran
6-Chloro-3-methylbenzofuran
Formel: C₉H₇ClO
Molecular Weight: 166,604 g/mol
Siedepunkt: 218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD15144093
CAS-Nummer: 30343-35-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR