2-Bromo-7-methoxyphenazine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR60266-1G
OR60266-250MG
OR60266-5G
APOSOR60266-1GEA
198
EUR
APOSOR60266-1G
APOSOR60266-250MG
APOSOR60266-5G
2-Bromo-7-methoxyphenazine 95%
2-Bromo-7-methoxyphenazine
Formel:
C₁₃H₉BrN₂O Schmelzpunkt: 174…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD32066843 CAS-Nummer: 58476-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...