5-Bromo-2-[(tetrahydro-2H-pyran-4-yl)oxy]pyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-((tetrahydro-2H-pyran-4-yl)oxy)pyridine, 4-[(5-Bromopyridin-2-yl)oxy]tetrahydro-2H-pyran, 4-[(5-Bromopyridin-2-yl)oxy]oxane

OR6070-1G OR6070-5G
APOSOR6070-1GEA 246 EUR
APOSOR6070-1G APOSOR6070-5G
5-Bromo-2-[(tetrahydro-2H-pyran-4-yl)oxy]pyridine 97%
5-Bromo-2-[(tetrahydro-2H-pyran-4-yl)oxy]pyridine
Formel: C₁₀H₁₂BrNO₂
Molecular Weight: 258.112 g/mol
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09064985
CAS-Nummer: 494772-07-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR