6-Methoxyisatoic anhydride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methoxy-2H-3,1-benzoxazine-2,4(1H)-dione, 5-Methoxy-1H-benzo[d][1,3]oxazine-2,4-dione
OR61001-5G
OR61001-250MG
OR61001-1G
APOSOR61001-5GEA
206
EUR
APOSOR61001-5G
APOSOR61001-250MG
APOSOR61001-1G
6-Methoxyisatoic anhydride
6-Methoxyisatoic anhydride
Formel:
C₉H₇NO₄ Molecular Weight: 193,156 g/mol Schmelzpunkt: 258…264 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11046419 CAS-Nummer: 67765-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...