4-Chloro-2-methyl-5-nitrobenzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Carboxy-5-chloro-4-nitrotoluene, 4-Chloro-5-nitro-o-toluic acid
OR61008-1G
OR61008-250MG
APOSOR61008-1GEA
126
EUR
APOSOR61008-1G
APOSOR61008-250MG
4-Chloro-2-methyl-5-nitrobenzoic acid 97%
4-Chloro-2-methyl-5-nitrobenzoic acid
Formel:
C₈H₆ClNO₄ Molecular Weight: 215,591 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071079 CAS-Nummer: 476660-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...