N-(Toluene-4-sulphonyl)glycine methyl ester
Lieferant: Apollo Scientific
Synonyme: Methyl N-[(4-methylphenyl)sulphonyl]glycinate, Tos-Gly-Ome, N-Tosylglycine methyl ester, Methyl [(4-methylphenyl)sulphonamido]acetate
OR61063-100G OR61063-25G OR61063-500G
APOSOR61063-100GEA 115 EUR
APOSOR61063-100G APOSOR61063-25G APOSOR61063-500G
N-(Toluene-4-sulphonyl)glycine methyl ester
N-(Toluene-4-sulphonyl)glycine methyl ester
Formel: C₁₀H₁₃NO₄S
Molecular Weight: 243.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00558890
CAS-Nummer: 2645-02-5