4-[2-(1H-Pyrazol-1-yl)ethoxy]benzeneboronic acid, pinacol ester 96%
Lieferant: Apollo Scientific
OR6107-1G
OR6107-250MG
APOSOR6107-1GEA
315
EUR
APOSOR6107-1G
APOSOR6107-250MG
4-[2-(1H-Pyrazol-1-yl)ethoxy]benzeneboronic acid, pinacol ester 96%
4-[2-(1H-Pyrazol-1-yl)ethoxy]benzeneboronic acid, pinacol ester
Formel:
C₁₇H₂₃BN₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258658 CAS-Nummer: 957061-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...