2',4',5'-Trichloroacetophenone 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2,4,5-Trichlorophenyl)ethanone
OR61087-100MG
OR61087-1G
OR61087-250MG
APOSOR61087-100MGEA
21.9
EUR
APOSOR61087-100MG
APOSOR61087-1G
APOSOR61087-250MG
2',4',5'-Trichloroacetophenone 95+%
2',4',5'-Trichloroacetophenone
Formel:
C₈H₅Cl₃O Molecular Weight: 223.484 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09261422 CAS-Nummer: 13061-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...