Methyl 3-bromo-2,4,6-trimethylbenzoate 95%

Lieferant: Apollo Scientific
Achtung

OR61097-1G OR61097-250MG
APOSOR61097-1GEA 647 EUR
APOSOR61097-1G APOSOR61097-250MG
Methyl 3-bromo-2,4,6-trimethylbenzoate 95%
Methyl 3-bromo-2,4,6-trimethylbenzoate
Formel: C₁₁H₁₃BrO₂
Molecular Weight: 257.124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00184978
CAS-Nummer: 26584-20-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR