1-(4-Bromophenyl)-3-cyano-1,2-dihydro-2-oxopyridine 97%
Lieferant: Apollo Scientific
Synonyme:
1-(4-Bromophenyl)-1,2-dihydro-2-oxopyridine-3-carbonitrile
OR6110-1G
OR6110-250MG
APOSOR6110-1GEA
517
EUR
APOSOR6110-1G
APOSOR6110-250MG
1-(4-Bromophenyl)-3-cyano-1,2-dihydro-2-oxopyridine 97%
1-(4-Bromophenyl)-3-cyano-1,2-dihydro-2-oxopyridine
Formel:
C₁₂H₇BrN₂O Molecular Weight: 275.101 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258784 CAS-Nummer: 929000-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...