3-Amino-3-phenyloxetane hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Aminooxetan-3-yl)benzene hydrochloride, 3-Phenyloxetan-3-amine hydrochloride

OR61143-1G OR61143-250MG
APOSOR61143-1GEA 830 EUR
APOSOR61143-1G APOSOR61143-250MG
3-Amino-3-phenyloxetane hydrochloride
3-Amino-3-phenyloxetane hydrochloride
Formel: C₉H₁₁NO·ClH
Molecular Weight: 185,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17215843
CAS-Nummer: 1211284-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR