2-Amino-7-bromo-1,3-benzothiazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-1,3-benzothiazol-2-amine, 7-Bromobenzo[d]thiazol-2-amine

OR61151-250MG
APOSOR61151-250MGEA 0 EUR
APOSOR61151-250MG
2-Amino-7-bromo-1,3-benzothiazole
2-Amino-7-bromo-1,3-benzothiazole
Formel: C₇H₅BrN₂S
Molecular Weight: 229.097 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11110957
CAS-Nummer: 20358-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR