5-Bromo-1H-indazole-7-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-7-carboxy-1H-indazole
OR61153-1G
OR61153-250MG
APOSOR61153-1GEA
265
EUR
APOSOR61153-1G
APOSOR61153-250MG
5-Bromo-1H-indazole-7-carboxylic acid
5-Bromo-1H-indazole-7-carboxylic acid
Formel:
C₈H₅BrN₂O₂ Molecular Weight: 241,042 g/mol Schmelzpunkt: 280…284 °C (Subl.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD13177019 CAS-Nummer: 953409-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...