5-Chloro-2-methoxyphenylacetic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Carboxymethyl)-4-chloroanisole, (5-Chloro-2-methoxyphenyl)acetic Acid

OR61292-1G OR61292-25G OR61292-5G
APOSOR61292-1GEA 23.2 EUR
APOSOR61292-1G APOSOR61292-25G APOSOR61292-5G
5-Chloro-2-methoxyphenylacetic acid 97%
5-Chloro-2-methoxyphenylacetic acid
Formel: C₉H₉ClO₃
Molecular Weight: 200,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664695
CAS-Nummer: 7569-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR