4-Amino-6-chloro-3H-imidazo[4,5-c]pyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-3H-imidazo[4,5-c]pyridin-4-amine
OR61299-100MG
OR61299-250MG
OR61299-1G
APOSOR61299-100MGEA
177
EUR
APOSOR61299-100MG
APOSOR61299-250MG
APOSOR61299-1G
4-Amino-6-chloro-3H-imidazo[4,5-c]pyridine 97%
4-Amino-6-chloro-3H-imidazo[4,5-c]pyridine
Formel:
C₆H₅ClN₄ Molecular Weight: 168,584 g/mol Schmelzpunkt: 266 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD18803412 CAS-Nummer: 52559-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...