5-Acetyl-8-(benzyloxy)quinolin-2(1H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[8-(Benzyloxy)-1,2-dihydro-2-oxoquinolin-5-yl]ethan-1-one, 5-Acetyl-8-(benzyloxy)-1,2-dihydro-2-oxoquinoline

OR61366-1G OR61366-500MG
APOSOR61366-1GEA 212 EUR
APOSOR61366-1G APOSOR61366-500MG
5-Acetyl-8-(benzyloxy)quinolin-2(1H)-one
5-Acetyl-8-(benzyloxy)quinolin-2(1H)-one
Formel: C₁₈H₁₅NO₃
Molecular Weight: 293,317 g/mol
Schmelzpunkt: 179…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD19381743
CAS-Nummer: 93609-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR