4-Iodo-1H-benzimidazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR61441-1G
OR61441-250MG
APOSOR61441-1GEA
377
EUR
APOSOR61441-1G
APOSOR61441-250MG
4-Iodo-1H-benzimidazole 95%
4-Iodo-1H-benzimidazole
Formel:
C₇H₅IN₂ Molecular Weight: 244,032 g/mol Schmelzpunkt: 188…192 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09910322 CAS-Nummer: 51288-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...