2-Bromo-5-iodobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromo-5-iodophenyl)methanol

OR61452-5G OR61452-25G
APOSOR61452-5GEA 21.9 EUR
APOSOR61452-5G APOSOR61452-25G
2-Bromo-5-iodobenzyl alcohol
2-Bromo-5-iodobenzyl alcohol
Formel: C₇H₆BrIO
Molecular Weight: 312,93 g/mol
Schmelzpunkt: 112…116 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11044842
CAS-Nummer: 946525-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR