5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-methyl-1H-imidazo[4,5-b]pyridine, 6-Bromo-2-methyl-7-azabenzimidazole

OR61523-1G OR61523-250MG
APOSOR61523-1GEA 198 EUR
APOSOR61523-1G APOSOR61523-250MG
5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
5-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
Formel: C₇H₆BrN₃
Molecular Weight: 212.047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16657201
CAS-Nummer: 219762-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR