3-Bromo-N-cyclopropyl-2-nitroaniline

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(3-Bromo-2-nitrophenyl)cyclopropylamine, 2-Bromo-6-(cyclopropylamino)nitrobenzene

OR61608-1G OR61608-250MG
APOSOR61608-1GEA 429 EUR
APOSOR61608-1G APOSOR61608-250MG
3-Bromo-N-cyclopropyl-2-nitroaniline
3-Bromo-N-cyclopropyl-2-nitroaniline
Formel: C₉H₉BrN₂O₂
Molecular Weight: 257,084 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22415284
CAS-Nummer: 1396554-46-3
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR