2-(4-Iodopyrazol-1-yl)acetic acid

Lieferant: Apollo Scientific
Achtung

OR62028-1G OR62028-250MG OR62028-5G
APOSOR62028-1GEA 49.3 EUR
APOSOR62028-1G APOSOR62028-250MG APOSOR62028-5G
2-(4-Iodopyrazol-1-yl)acetic acid
2-(4-Iodopyrazol-1-yl)acetic acid
Formel: C₅H₅IN₂O₂
Molecular Weight: 252,01 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08558964
CAS-Nummer: 6752-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR