1-(4-Iodopyrazol-1-yl)propan-2-one

Lieferant: Apollo Scientific
Achtung

OR62030-1G OR62030-250MG OR62030-5G
APOSOR62030-1GEA 522 EUR
APOSOR62030-1G APOSOR62030-250MG APOSOR62030-5G
1-(4-Iodopyrazol-1-yl)propan-2-one
1-(4-Iodopyrazol-1-yl)propan-2-one
Formel: C₆H₇IN₂O
Molecular Weight: 250.037 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20327030
CAS-Nummer: 1343366-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR