Ethyl 3-(4-iodopyrazol-1-yl)propanoate
Lieferant: Apollo Scientific
|
Achtung
|
OR62033-1G
OR62033-250MG
APOSOR62033-1GEA
59.6
EUR
APOSOR62033-1G
APOSOR62033-250MG
Ethyl 3-(4-iodopyrazol-1-yl)propanoate
Ethyl 3-(4-iodopyrazol-1-yl)propanoate
Formel:
C₈H₁₁IN₂O₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD20326732 CAS-Nummer: 6645-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...