Sodium (1H-indol-2-yl)acetate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1H-Indol-2-yl)acetic acid sodium salt, Sodium 2-(1H-indol-2-yl)ethanoate
OR6306A-100MG
OR6306A-250MG
OR6306A-25MG
APOSOR6306A-100MGEA
109
EUR
APOSOR6306A-100MG
APOSOR6306A-250MG
APOSOR6306A-25MG
Sodium (1H-indol-2-yl)acetate 95%
Sodium (1H-indol-2-yl)acetate
Formel:
C₁₀H₈NO₂·Na Molecular Weight: 197,166 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09787506 CAS-Nummer: 172513-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...