3-(4-Aminophenyl)-4-bromo-1-methylpyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Bromo-1-methyl-1H-pyrazol-3-yl)aniline

OR6359-1G OR6359-500MG
APOSOR6359-1GEA 379 EUR
APOSOR6359-1G APOSOR6359-500MG
3-(4-Aminophenyl)-4-bromo-1-methylpyrazole
3-(4-Aminophenyl)-4-bromo-1-methylpyrazole
Formel: C₁₀H₁₀BrN₃
Molecular Weight: 252,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00084884
CAS-Nummer: 175276-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR