1-(2-Hydroxy-5-iodo-3-methylbenzyl)-1,3,3-trimethylpropanediamine dihydrochloride

Lieferant: Apollo Scientific
Achtung

OR6460-100MG OR6460-200MG
APOSOR6460-100MGEA 279 EUR
APOSOR6460-100MG APOSOR6460-200MG
1-(2-Hydroxy-5-iodo-3-methylbenzyl)-1,3,3-trimethylpropanediamine dihydrochloride
1-(2-Hydroxy-5-iodo-3-methylbenzyl)-1,3,3-trimethylpropanediamine dihydrochloride

Radiopharmaceutical precursor

Formel: C₁₄H₂₃IN₂O·2ClH
Molecular Weight: 435,172 g/mol
Schmelzpunkt: 160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040808
CAS-Nummer: 89815-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR