3-Bromocinnamic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-Bromophenyl)acrylic acid, 3-(3-Bromophenyl)prop-2-enoic acid
OR6579-100G
APOSOR6579-100GEA
85.7
EUR
APOSOR6579-100G
3-Bromocinnamic acid 97%
3-Bromocinnamic acid
Formel:
C₉H₇BrO₂ Molecular Weight: 227,055 g/mol Schmelzpunkt: 177…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004382 CAS-Nummer: 32862-97-8 EINECS: 251-267-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...