N1-Benzylbenzene-1,2-diamine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N1-(Phenylmethyl)benzene-1,2-diamine, N1-Benzylphenylene-1,2-diamine
OR6664-5G
OR6664-10G
APOSOR6664-5GEA
105
EUR
APOSOR6664-5G
APOSOR6664-10G
N1-Benzylbenzene-1,2-diamine 95%
N1-Benzylbenzene-1,2-diamine
Formel:
C₁₃H₁₄N₂ Molecular Weight: 198,264 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094682 CAS-Nummer: 5822-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...