4-Dimethylamino-2-hydroxybenzaldehyde 98%
Lieferant: Apollo Scientific
Synonyme:
4-(Dimethylamino)-2-hydroxybenzaldehyde, 4-(Dimethylamino)salicylaldehyde, 5-(Dimethylamino)-2-formylphenol
OR6690-5G
APOSOR6690-5GEA
142
EUR
APOSOR6690-5G
4-Dimethylamino-2-hydroxybenzaldehyde 98%
4-Dimethylamino-2-hydroxybenzaldehyde
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,189 g/mol Schmelzpunkt: 78 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00027419 CAS-Nummer: 41602-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...