5-Amino-3-(fur-2-yl)isoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Furan-2-yl)isoxazol-5-amine

OR6697-150MG OR6697-50MG
APOSOR6697-150MGEA 147 EUR
APOSOR6697-150MG APOSOR6697-50MG
5-Amino-3-(fur-2-yl)isoxazole
5-Amino-3-(fur-2-yl)isoxazole
Formel: C₇H₆N₂O₂
Molecular Weight: 150.135 g/mol
Schmelzpunkt: 99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01754641
CAS-Nummer: 33866-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR