[3-(Pyrrolidin-1-yl)phenyl]methylamine 90%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(Pyrrolidin-1-yl)benzylamine, 1-[3-(Aminomethyl)phenyl]pyrrolidine

OR6732-1G OR6732-250MG
APOSOR6732-1GEA 328 EUR
APOSOR6732-1G APOSOR6732-250MG
[3-(Pyrrolidin-1-yl)phenyl]methylamine 90%
[3-(Pyrrolidin-1-yl)phenyl]methylamine
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772827
CAS-Nummer: 175696-70-5
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR