3-(Pyrrolidin-1-yl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

OR6744-1G OR6744-5G
APOSOR6744-1GEA 143 EUR
APOSOR6744-1G APOSOR6744-5G
3-(Pyrrolidin-1-yl)benzoic acid 97%
3-(Pyrrolidin-1-yl)benzoic acid
Formel: C₁₁H₁₃NO₂
Schmelzpunkt: 215…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426724
CAS-Nummer: 72548-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR