2,5-Dimethyl-4-(hydroxymethyl)-1,3-oxazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2,5-Dimethyl-1,3-oxazol-4-yl)methanol, (2,5-Dimethyloxazol-4-yl)methanol
OR6751-1G
OR6751-250MG
OR6751-5G
APOSOR6751-1GEA
139
EUR
APOSOR6751-1G
APOSOR6751-250MG
APOSOR6751-5G
2,5-Dimethyl-4-(hydroxymethyl)-1,3-oxazole 97%
2,5-Dimethyl-4-(hydroxymethyl)-1,3-oxazole
Formel:
C₆H₉NO₂ Schmelzpunkt: 73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07772804 CAS-Nummer: 92901-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...