N-Methyl-1-[2-(pyrrolidin-1-yl)pyridin-3-yl]methylamine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-{3-[(Methylamino)methyl]pyridin-2-yl}pyrrolidine, 3-[(Methylamino)methyl]-2-(pyrrolidin-1-yl)pyridine
OR6796-1G
OR6796-250MG
APOSOR6796-1GEA
394
EUR
APOSOR6796-1G
APOSOR6796-250MG
N-Methyl-1-[2-(pyrrolidin-1-yl)pyridin-3-yl]methylamine 97%
N-Methyl-1-[2-(pyrrolidin-1-yl)pyridin-3-yl]methylamine
Formel:
C₁₁H₁₇N₃ Molecular Weight: 191,273 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07772854 CAS-Nummer: 857284-17-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...