N-Methyl-3-(pyrimidin-5-yl)benzylamine 97%

Lieferant: Apollo Scientific

OR6800-1G OR6800-250MG
APOSOR6800-1GEA 893 EUR
APOSOR6800-1G APOSOR6800-250MG
N-Methyl-3-(pyrimidin-5-yl)benzylamine 97%
N-Methyl-3-(pyrimidin-5-yl)benzylamine
Formel: C₁₂H₁₃N₃
Molecular Weight: 199,252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08060515
CAS-Nummer: 852431-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR