4-Chloro-6-methoxy-1H-indole 95+%

Lieferant: Apollo Scientific

OR6800T-100MG OR6800T-250MG OR6800T-1G
APOSOR6800T-100MGEA 102 EUR
APOSOR6800T-100MG APOSOR6800T-250MG APOSOR6800T-1G
4-Chloro-6-methoxy-1H-indole 95+%
4-Chloro-6-methoxy-1H-indole

A promutagen derived from fava beans.

Formel: C₉H₈ClNO
Molecular Weight: 181,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00238559
CAS-Nummer: 93490-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR