{3-[2-(Morpholin-4-yl)ethoxy]phenyl}methylamine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-{2-[3-(Aminomethyl)phenoxy]ethyl}morpholine, 3-[2-(Morpholin-4-yl)ethoxy]benzylamine

OR6807-1G OR6807-250MG
APOSOR6807-1GEA 0 EUR
APOSOR6807-1G APOSOR6807-250MG
{3-[2-(Morpholin-4-yl)ethoxy]phenyl}methylamine 95%
{3-[2-(Morpholin-4-yl)ethoxy]phenyl}methylamine
Formel: C₁₃H₂₀N₂O₂
Molecular Weight: 236,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772847
CAS-Nummer: 857284-08-3
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR