1-[4-(Benzyloxy)phenyl]piperazine hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzyl 4-(piperazin-1-yl)phenyl ether hydrochloride
OR6844-1G
OR6844-2G
OR6844-500MG
APOSOR6844-1GEA
232
EUR
APOSOR6844-1G
APOSOR6844-2G
APOSOR6844-500MG
1-[4-(Benzyloxy)phenyl]piperazine hydrochloride 97%
1-[4-(Benzyloxy)phenyl]piperazine hydrochloride
Formel:
C₁₇H₂₀N₂O·ClH Molecular Weight: 304,814 g/mol Schmelzpunkt: 210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191210 CAS-Nummer: 321132-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...