1-(2-Diallylaminoethyl)piperazine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: N-[2-(Piperazin-1-yl)ethyl]-N-(prop-2-en-1-yl)prop-2-en-1-amine

OR6911-1G OR6911-500MG OR6911-5G
APOSOR6911-1GEA 111 EUR
APOSOR6911-1G APOSOR6911-500MG APOSOR6911-5G
1-(2-Diallylaminoethyl)piperazine 97%
1-(2-Diallylaminoethyl)piperazine
Formel: C₁₂H₂₃N₃
Molecular Weight: 209.331 g/mol
Siedepunkt: 76…78 °C (0.5 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00191216
CAS-Nummer: 199475-35-9
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR