Ethyl 1-benzylpiperidine-3-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

OR6928-5G OR6928-1G OR6928-25G
APOSOR6928-5GEA 68.4 EUR
APOSOR6928-5G APOSOR6928-1G APOSOR6928-25G
Ethyl 1-benzylpiperidine-3-carboxylate 98%
Ethyl 1-benzylpiperidine-3-carboxylate
Formel: C₁₅H₂₁NO₂
Molecular Weight: 247,333 g/mol
Siedepunkt: 125…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191221
CAS-Nummer: 72551-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR