5-Bromothiophene-3-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-4-formylthiophene, 5-Bromothiophene-3-carbaldehyde, 5-Bromo-3-thenaldehyde

OR70005-1G OR70005-250MG OR70005-5G
APOSOR70005-1GEA 91.5 EUR
APOSOR70005-1G APOSOR70005-250MG APOSOR70005-5G
5-Bromothiophene-3-carboxaldehyde 97%
5-Bromothiophene-3-carboxaldehyde
Formel: C₅H₃BrOS
Molecular Weight: 191.046 g/mol
Siedepunkt: 114…116 °C (17 mm Hg)
Lagertemperatur: Kühlschrank
MDL: MFCD09042555
CAS-Nummer: 18791-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR